Sodium benzoate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Benzoic acid sodium salt

M-Clarity™ quality level = MQ200

B3420-1KG B3420-250G
SIALB3420-1KGEA 122 EUR
SIALB3420-1KG SIALB3420-250G
Sodium benzoate, Sigma-Aldrich®
Sodium benzoate
Formula: C₆H₅COONa
Melting Pt: 410…430 °C
Density: 1.44 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 13,08654

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR