3-Aminobenzoic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
127671-100G 127671-25G 127671-500G
SIAL127671-100GEA 52.2 EUR
SIAL127671-100G SIAL127671-25G SIAL127671-500G
3-Aminobenzoic acid, Sigma-Aldrich®
3-Aminobenzoic acid
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 308
Melting Pt: 174
Density: 1.5105 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007795
CAS Number: 99-05-8
EINECS: 202-724-4
Merck Index: 13,00420