3-Amino-p-anisic acid
Supplier: Thermo Scientific
Warning
SB00931.1 SB00931.10 SB00931.50
MAYBSB00931.1EA 0 EUR
MAYBSB00931.1 MAYBSB00931.10 MAYBSB00931.50
3-Amino-p-anisic acid
3-Amino-p-anisic acid
Formula: C₈H₉NO₃
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 204 °C
Storage Temperature: Ambient
MDL Number: MFCD00002521
CAS Number: 2840-26-8
EINECS: 220-634-3