3-Aminosalicylic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
3-Aminosalicylic Acid, 3-Amino-2-hydroxybenzoic acid, 3-Amino-2-hydroxybenzoic acid
M-Clarity™ quality level = MQ200
255300-500MG
255300-100MG
SIAL255300-500MGEA
253
EUR
SIAL255300-500MG
SIAL255300-100MG
3-Aminosalicylic acid, Sigma-Aldrich®
3-Aminosalicylic acid
Formula:
C₇H₇NO₃ MW: 153.14 g/mol Boiling Pt: 347 °C (1013 hPa) Melting Pt: 290 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010299 CAS Number: 570-23-0 EINECS: 209-328-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...