3-Amino-4-hydroxybenzoic acid, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
289647-25G 289647-5G
SIAL289647-25GEA 87.5 EUR
SIAL289647-25G SIAL289647-5G
3-Amino-4-hydroxybenzoic acid, Sigma-Aldrich®
3-Amino-4-hydroxybenzoic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 201 °C
Storage Temperature: Ambient
MDL Number: MFCD00007697
CAS Number: 1571-72-8
EINECS: 216-390-2