5-Aminosalicylic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
A3537-100G A3537-25G
SIALA3537-100GEA 593 EUR
SIALA3537-100G SIALA3537-25G
5-Aminosalicylic acid, Sigma-Aldrich®
5-Aminosalicylic acid
Formula: C₇H₇NO₃
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 280 °C
Density: 1.568 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007877
CAS Number: 89-57-6
EINECS: 201-919-1
Merck Index: 12,05964