4-Aminosalicylic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 4-Amino-2-hydroxybenzoic acid
M-Clarity™ quality level = MQ200
A79604-100G A79604-5G
SIALA79604-100GEA 63.1 EUR
SIALA79604-100G SIALA79604-5G
4-Aminosalicylic acid, Sigma-Aldrich®
4-Aminosalicylic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 150 °C
Density: 1.545 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007789
CAS Number: 65-49-6
EINECS: 200-613-5
Merck Index: 13,00476