4-Amino-3-hydroxybenzoic acid, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

339598-25G 339598-5G
SIAL339598-25GEA 122 EUR
SIAL339598-25G SIAL339598-5G
4-Amino-3-hydroxybenzoic acid, Sigma-Aldrich®
4-Amino-3-hydroxybenzoic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017094
CAS Number: 2374-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR