2-Nitrobenzoic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
127698-100G 127698-5G
SIAL127698-100GEA 125 EUR
SIAL127698-100G SIAL127698-5G
2-Nitrobenzoic acid, Sigma-Aldrich®
2-Nitrobenzoic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Melting Pt: 146…148 °C
Density: 1.575 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007137
CAS Number: 552-16-9
EINECS: 209-004-9
Merck Index: 12,06686