3-Nitrobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: m-Nitrobenzoic acid
M-Clarity™ quality level = MQ200
185329-100G 185329-500G 185329-5G
SIAL185329-100GEA 41.3 EUR
SIAL185329-100G SIAL185329-500G SIAL185329-5G
3-Nitrobenzoic acid, Sigma-Aldrich®
3-Nitrobenzoic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 341
Melting Pt: 139…142 °C
Density: 1.494 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007251
CAS Number: 121-92-6
EINECS: 204-508-5
Merck Index: 12,06686