3-Hydroxy-4-nitrobenzoic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
H48409-100G
H48409-25G
SIALH48409-100GEA
295
EUR
SIALH48409-100G
SIALH48409-25G
3-Hydroxy-4-nitrobenzoic acid, Sigma-Aldrich®
3-Hydroxy-4-nitrobenzoic acid
Formula:
C₇H₅NO₅ MW: 183.12 g/mol Melting Pt: 234…235 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007110 CAS Number: 619-14-7 EINECS: 210-580-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...