4-Hydroxy-3-nitrobenzoic acid, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

228575-100G 228575-25G
SIAL228575-100GEA 227 EUR
SIAL228575-100G SIAL228575-25G
4-Hydroxy-3-nitrobenzoic acid, Sigma-Aldrich®
4-Hydroxy-3-nitrobenzoic acid
Formula: C₇H₅NO₅
MW: 183.12 g/mol
Melting Pt: 182…185 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007119
CAS Number: 616-82-0
EINECS: 210-494-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR