3-Cyclopentyloxy-4-methoxybenzoic acid

Supplier: Thermo Scientific

KM09079DA KM09079EA
MAYBKM09079DAEA 0 EUR
MAYBKM09079DA MAYBKM09079EA
3-Cyclopentyloxy-4-methoxybenzoic acid
3-Cyclopentyloxy-4-methoxybenzoic acid
Formula: C₁₃H₁₆O₄
MW: 236.27 g/mol
CAS Number: 144036-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR