3,4,5-Trimethoxybenzoic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Trimethylgallic Acid
M-Clarity™ quality level = MQ100
T69000-100G
T69000-5G
SIALT69000-100GEA
57.2
EUR
SIALT69000-100G
SIALT69000-5G
3,4,5-Trimethoxybenzoic acid, Sigma-Aldrich®
3,4,5-Trimethoxybenzoic acid
Formula:
C₁₀H₁₂O₅ MW: 212.2 g/mol Boiling Pt: 376 °C (1013 hPa) Melting Pt: 168…170 °C Density: 1.31 g/cm³ (24.95 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002501 CAS Number: 118-41-2 EINECS: 204-248-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...