Salicylic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
SIAL105910-3KGEA 158 EUR
SIAL105910-3KG SIAL105910-500G
Salicylic acid, Sigma-Aldrich®
Salicylic acid
Formula: (HO)C₆H₄CO₂H
MW: 138,12 g/mol
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 157…159 °C
Density: 1,443 g/cm³ (20 °C)
Flash Pt: 157 °C
MDL Number: MFCD00002439
CAS Number: 69-72-7
Merck Index: 13,08411
https://si.vwr.com/stibo/web/std.lang.all/99/00/749900.jpg