Salicylic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIALS7401-500GEA 65 EUR
SIALS7401-500G
Salicylic acid, Sigma-Aldrich®
Salicylic acid
Formula: (HO)C₆H₄CO₂H
MW: 138,12 g/mol
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 157…159 °C
Density: 1,443 g/cm³ (20 °C)
Flash Pt: 157 °C
MDL Number: MFCD00002439
CAS Number: 69-72-7
Merck Index: 12,08484
https://si.vwr.com/stibo/web/std.lang.all/99/00/749900.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR