Sodium salicylate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Sodium 2-hydroxybenzoate, 2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt
M-Clarity™ quality level = MQ200
S2679-100G S2679-1KG S2679-500G
SIALS2679-100GEA 66.4 EUR
SIALS2679-100G SIALS2679-1KG SIALS2679-500G
Sodium salicylate, Sigma-Aldrich®
Sodium salicylate
Formula: HOC₆H₄COONa
MW: 160.10 g/mol
Melting Pt: 200
Density: 0.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002440
CAS Number: 54-21-7
EINECS: 200-198-0
Merck Index: 12,08819