2,6-Dihydroxybenzoic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
D109606-100G
D109606-25G
SIALD109606-100GEA
53
EUR
SIALD109606-100G
SIALD109606-25G
2,6-Dihydroxybenzoic acid, Sigma-Aldrich®
2,6-Dihydroxybenzoic acid
Formula:
C₇H₆O₄ Boiling Pt: 339 °C (1013 hPa) Melting Pt: 152…158 °C Density: 1.58 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002462 CAS Number: 303-07-1 EINECS: 206-134-8 |