Gallic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
3,4,5-Trihydroxybenzoic acid
M-Clarity™ quality level = MQ200
G7384-100G
G7384-1KG
G7384-250G
SIALG7384-100GEA
91.2
EUR
SIALG7384-100G
SIALG7384-1KG
SIALG7384-250G
Gallic acid, Sigma-Aldrich®
Gallic acid
Formula:
C₇H₆O₅ MW: 170.12 g/mol Boiling Pt: 501 °C (1013 hPa) Melting Pt: 251…252 °C Density: 1.694 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002510 CAS Number: 149-91-7 EINECS: 205-749-9 Merck Index: 13,04366 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...