2-(Methylthio)benzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Warning
A10379.14 A10379.06
A10379.14EA 307 EUR
A10379.14 A10379.06
2-(Methylthio)benzoic acid ≥98%
2-(Methylthio)benzoic acid
Purity: 98+% CAS No.: 3724-10-5 Molecular Formula: C8H8O2S Formula Weight: 168.21 Melting Point: 166-168° MDL No.: MFCD00029934
Formula: C₈H₈O₂S
MW: 168.22 g/mol
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00029934
CAS Number: 3724-10-5