Thiosalicylic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
2-Thiobenzoic acid, 2-Mercaptobenzoic acid
M-Clarity™ quality level = MQ100
T33200-100G
T33200-500G
T33200-5G
SIALT33200-100GEA
62.1
EUR
SIALT33200-100G
SIALT33200-500G
SIALT33200-5G
Thiosalicylic acid, Sigma-Aldrich®
Thiosalicylic acid
Formula:
C₇H₆O₂S MW: 154.19 g/mol Boiling Pt: 230 °C (0.02 mmHg) Melting Pt: 165…166 °C Density: 1.489 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004836 CAS Number: 147-93-3 EINECS: 205-704-3 UN: 3335 ADR: 9, Merck Index: 12,09498 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...