Trimellitic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1,2,4-Benzenetricarboxylic acid, Benzene-1,2,4-tricarboxylic acid

M-Clarity™ quality level = MQ200

B4589-100G B4589-5G
SIALB4589-100GEA 189 EUR
SIALB4589-100G SIALB4589-5G
Trimellitic acid, Sigma-Aldrich®
Trimellitic acid
Formula: C₉H₆O₆
MW: 210.14 g/mol
Boiling Pt: 107…113 °C (0.1 mmHg)
Melting Pt: 240…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00002470
CAS Number: 528-44-9
EINECS: 208-432-3
Merck Index: 12,09832

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR