Phenyl salicylate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Phenyl 2-hydroxybenzoate, Phenyl Salicylate
M-Clarity™ quality level = MQ200
149187-250G 149187-50G
SIAL149187-250GEA 82.8 EUR
SIAL149187-250G SIAL149187-50G
Phenyl salicylate, Sigma-Aldrich®
Phenyl salicylate
Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Boiling Pt: 307 °C (1013 hPa)
Melting Pt: 41…43 °C
Density: 1.307 g/cm³ (20 °C)
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00002213
CAS Number: 118-55-8
EINECS: 204-259-2
UN: 3077
ADR: 9,III
Merck Index: 12,07464