Phenyl salicylate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Phenyl 2-hydroxybenzoate, Phenyl Salicylate
M-Clarity™ quality level = MQ300
W396001-1KG W396001-5KG W396001-SAMPLE
SIALW396001-1KGEA 132 EUR
SIALW396001-1KG SIALW396001-5KG SIALW396001-SAMPLE
Phenyl salicylate, Sigma-Aldrich®
Phenyl salicylate
Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Boiling Pt: 307 °C (1013 hPa)
Melting Pt: 41…43 °C
Density: 1.307 g/cm³ (20 °C)
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00002213
CAS Number: 118-55-8
EINECS: 204-259-2
UN: 3077
ADR: 9,III
Merck Index: 12,07464