1,2,4-Trifluorobenzene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
196843-5G 196843-25G
SIAL196843-5GEA 24.7 EUR
SIAL196843-5G SIAL196843-25G
1,2,4-Trifluorobenzene, Sigma-Aldrich®
1,2,4-Trifluorobenzene
Formula: C₆H₃F₃
MW: 132.09 g/mol
Boiling Pt: 88 °C (1013 hPa)
Melting Pt: –35 °C
Density: 1.26 g/cm³ (20 °C)
MDL Number: MFCD00000305
CAS Number: 367-23-7
EINECS: 206-684-9
UN: 2387
ADR: 3,II