Fluorobenzene, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Phenylfluoride
M-Clarity™ quality level = MQ200
F6001-100G-A
F6001-500G-A
F6001-5G-A
SIALF6001-100G-AEA
42.4
EUR
SIALF6001-100G-A
SIALF6001-500G-A
SIALF6001-5G-A
Fluorobenzene, Sigma-Aldrich®
Fluorobenzene
Formula:
C₆H₅F MW: 96.1 g/mol Boiling Pt: 85.1 °C (1013 hPa) Melting Pt: –41 °C Density: 1.023 g/cm³ (20 °C) Flash Pt: –15 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000280 CAS Number: 462-06-6 EINECS: 207-321-7 UN: 2387 ADR: 3,II Merck Index: 13,04197 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...