2,3,4,5-Tetrafluorophenylboronic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2,3,4,5-Tetrafluorobenzeneboronic acid, Dihydroxy(2,3,4,5-tetrafluorophenyl)borane, (2,3,4,5-Tetrafluorophenyl)metaboric acid, (2,3,4,5-Tetrafluorophenyl)boronic acid

M-Clarity™ quality level = MQ100

696587-1G 696587-250MG
SIAL696587-1GEA 243 EUR
SIAL696587-1G SIAL696587-250MG
2,3,4,5-Tetrafluorophenylboronic acid, Sigma-Aldrich®
2,3,4,5-Tetrafluorophenylboronic acid
Formula: C₆H₃BF₄O₂
MW: 193.89 g/mol
MDL Number: MFCD04039308
CAS Number: 179923-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR