2,4-Difluorophenylboronic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
465070-25G
465070-5G
SIAL465070-25GEA
231
EUR
SIAL465070-25G
SIAL465070-5G
2,4-Difluorophenylboronic acid, Sigma-Aldrich®
2,4-Difluorophenylboronic acid
Formula:
C₆H₅BF₂O₂ MW: 157.91 g/mol |
MDL Number:
MFCD01318998 CAS Number: 144025-03-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...