2-Fluorophenylboronic acid 98%
Supplier: Thermo Scientific
Warning
Synonyms: Dihydroxy(2-fluorophenyl)borane, 2-Fluorobenzeneboronic acid, (2-Fluorophenyl)metaboric acid, (2-Fluorophenyl)boronic acid
AC35934.1 AC35934.10 359340010 359340050
MAYBAC35934.1EA 0 EUR
MAYBAC35934.1 MAYBAC35934.10 ACRO359340010 ACRO359340050
2-Fluorophenylboronic acid 98%
2-Fluorophenylboronic acid
Formula: C₆H₆BFO₂
MW: 139.92 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00674013
CAS Number: 1993-03-9
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %