2,3,5-Trifluorobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
529192-1ML
529192-5ML
SIAL529192-1MLEA
48.7
EUR
SIAL529192-1ML
SIAL529192-5ML
2,3,5-Trifluorobenzaldehyde, Sigma-Aldrich®
2,3,5-Trifluorobenzaldehyde
Formula:
C₇H₃F₃O MW: 160.1 g/mol Boiling Pt: 165 °C (1013 hPa) Density: 1.407 g/cm³ (20 °C) |
MDL Number:
MFCD00083538 CAS Number: 126202-23-1 UN: 1989 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...