2,4-Difluorobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
265179-25G 265179-5G
SIAL265179-25GEA 82.9 EUR
SIAL265179-25G SIAL265179-5G
2,4-Difluorobenzaldehyde, Sigma-Aldrich®
2,4-Difluorobenzaldehyde
Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 82 °C (42 mmHg)
Density: 1.299 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010326
CAS Number: 1550-35-2
EINECS: 216-287-2
UN: 1989
ADR: 3 III