2,5-Difluorobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
265187-1G
265187-25G
265187-5G
SIAL265187-1GEA
27.7
EUR
SIAL265187-1G
SIAL265187-25G
SIAL265187-5G
2,5-Difluorobenzaldehyde, Sigma-Aldrich®
2,5-Difluorobenzaldehyde
Formula:
C₇H₄F₂O MW: 142.1 g/mol Boiling Pt: 65…66 °C (20 mmHg) Density: 1.3 g/cm³ (20 °C) |
MDL Number:
MFCD00010327 CAS Number: 2646-90-4 UN: 1989 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...