2-Fluorobenzaldehyde, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

F4807-100G F4807-10G
SIALF4807-100GEA 61.6 EUR
SIALF4807-100G SIALF4807-10G
2-Fluorobenzaldehyde, Sigma-Aldrich®
2-Fluorobenzaldehyde
Formula: C₇H₅FO
MW: 124.11 g/mol
Boiling Pt: 175 °C (1013 hPa)
Melting Pt: –44.5 °C
Density: 1.19 g/cm³ (20 °C)
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00003302
CAS Number: 446-52-6
EINECS: 207-171-2
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR