3-Fluorobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
F5005-10G
F5005-50G
SIALF5005-10GEA
43.2
EUR
SIALF5005-10G
SIALF5005-50G
3-Fluorobenzaldehyde, Sigma-Aldrich®
3-Fluorobenzaldehyde
Formula:
C₇H₅FO MW: 124.11 g/mol Boiling Pt: 172…174 °C (1013 hPa) Density: 1.7156 g/cm³ (20 °C) Flash Pt: 56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003348 CAS Number: 456-48-4 EINECS: 207-266-9 UN: 1989 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...