3-(Difluoromethoxy)benzaldehyde ≥97%
Supplier: Thermo Fisher Scientific
Warning
H26580.03 H26580.06
H26580.03EA 97.1 EUR
H26580.03 H26580.06
3-(Difluoromethoxy)benzaldehyde ≥97%
3-(Difluoromethoxy)benzaldehyde
Purity: 97% CAS No.: 85684-61-3 Molecular Formula: C8H6F2O2 Formula Weight: 172.13 MDL No.: MFCD00236222
Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236222
CAS Number: 85684-61-3
EINECS: None