3-Fluorobenzonitrile, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIAL235822-10GEA 28.1 EUR
SIAL235822-10G
3-Fluorobenzonitrile, Sigma-Aldrich®
3-Fluorobenzonitrile
Formula: C₇H₄FN
MW: 121,11 g/mol
Boiling Pt: 182 °C (1013 hPa)
Melting Pt: –16 °C
Density: 1,1334 g/cm³ (25 °C)
MDL Number: MFCD00001797
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR