4-Fluorobenzonitrile, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: p-Fluorobenzonitrile
M-Clarity™ quality level = MQ200
139416-25G 139416-5G
SIAL139416-25GEA 50.2 EUR
SIAL139416-25G SIAL139416-5G
4-Fluorobenzonitrile, Sigma-Aldrich®
4-Fluorobenzonitrile
Formula: C₇H₄FN
Boiling Pt: 188 °C (1013 hPa)
Melting Pt: 37…40 °C
Density: ∼1 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00001812
CAS Number: 1194-02-1
EINECS: 214-784-9
UN: 3439
ADR: 6.1,III