2,6-Difluorobenzoyl chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
249408-100G 249408-25G
SIAL249408-100GEA 198 EUR
SIAL249408-100G SIAL249408-25G
2,6-Difluorobenzoyl chloride, Sigma-Aldrich®
2,6-Difluorobenzoyl chloride
Formula: C₇H₃ClF₂O
Boiling Pt: 72 °C (1013 hPa)
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000659
CAS Number: 18063-02-0
EINECS: 241-971-2
UN: 3265
ADR: 8,II