3,5-Difluorobenzoyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
290254-25G 290254-5G
SIAL290254-25GEA 180 EUR
SIAL290254-25G SIAL290254-5G
3,5-Difluorobenzoyl chloride, Sigma-Aldrich®
3,5-Difluorobenzoyl chloride
Formula: C₇H₃ClF₂O
MW: 176.55 g/mol
Density: 1.5 g/cm³ (20 °C)
MDL Number: MFCD00010309
CAS Number: 129714-97-2
UN: 3265
ADR: 8 II