Pentafluorobenzoyl chloride 99%, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2,3,4,5,6-Pentafluorobenzoyl chloride, Pentafluorobenzoylchloride, PFBC
103772-1G 103772-25G 103772-5G
SIAL103772-1GEA 30.4 EUR
SIAL103772-1G SIAL103772-25G SIAL103772-5G
Pentafluorobenzoyl chloride 99%, Sigma-Aldrich®
Pentafluorobenzoyl chloride
Formula: C₇ClF₅O
MW: 230.52 g/mol
Boiling Pt: 158 °C (1013 hPa)
Density: 1.6 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000657
CAS Number: 2251-50-5
EINECS: 218-842-4
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance (Color) Colorless to Light Yellow
Appearance (Form) Liquid
Infrared spectrum Conforms to Structure
GC (area %) ≥98.5%