Ethyl-4-fluorobenzoate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Fluorobenzoic acid ethyl ester, Ethyl 4-fluorobenzoate
M-Clarity™ quality level = MQ100
102644-100G 102644-25G
SIAL102644-100GEA 97.2 EUR
SIAL102644-100G SIAL102644-25G
Ethyl-4-fluorobenzoate, Sigma-Aldrich®
Ethyl 4-fluorobenzoate
Formula: C₉H₉FO₂
MW: 168.17 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 26 °C
Density: 1.146 g/cm³ (20 °C)
MDL Number: MFCD00000351
CAS Number: 451-46-7
EINECS: 207-194-8
Merck Index: 13,04198