2,6-Difluorobenzoic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
190039-25G
190039-5G
SIAL190039-25GEA
106
EUR
SIAL190039-25G
SIAL190039-5G
2,6-Difluorobenzoic acid, Sigma-Aldrich®
2,6-Difluorobenzoic acid
Formula:
C₇H₄F₂O₂ MW: 158.1 g/mol Boiling Pt: 72…77 °C (13 mmHg) Melting Pt: 159…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002411 CAS Number: 385-00-2 EINECS: 206-856-3 |