2-Fluorobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: o-Fluorobenzoic acid
M-Clarity™ quality level = MQ100
412244-100G 412244-25G
SIAL412244-100GEA 73.5 EUR
SIAL412244-100G SIAL412244-25G
2-Fluorobenzoic acid, Sigma-Aldrich®
2-Fluorobenzoic acid
Formula: C₇H₅FO₂
MW: 140.11 g/mol
Boiling Pt: 245
Melting Pt: 121…124 °C
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002405
CAS Number: 445-29-4
EINECS: 207-158-1