4-Fluorobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
128384-5G 128384-25G
SIAL128384-5GEA 13.9 EUR
SIAL128384-5G SIAL128384-25G
4-Fluorobenzoic acid, Sigma-Aldrich®
4-Fluorobenzoic acid
Formula: C₇H₅FO₂
MW: 140.11 g/mol
Boiling Pt: 254
Melting Pt: 185
Density: 1.49 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002530
CAS Number: 456-22-4
EINECS: 207-259-0
Merck Index: 13,04198