4-Fluorobenzoic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
418846-1G
418846-5G
SIAL418846-1GEA
40.2
EUR
SIAL418846-1G
SIAL418846-5G
4-Fluorobenzoic acid, Sigma-Aldrich®
4-Fluorobenzoic acid
Formula:
C₇H₅FO₂ Boiling Pt: 254 °C (1013 hPa) Melting Pt: 185 °C Density: 1.49 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002530 CAS Number: 456-22-4 EINECS: 207-259-0 Merck Index: 13,04198 |