3-((Trifluoromethyl)thio)benzoic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

470090-1G 470090-250MG
SIAL470090-1GEA 126 EUR
SIAL470090-1G SIAL470090-250MG
3-((Trifluoromethyl)thio)benzoic acid, Sigma-Aldrich®
3-((Trifluoromethyl)thio)benzoic acid
Formula: C₈H₅F₃O₂S
MW: 222.19 g/mol
MDL Number: MFCD00236337
CAS Number: 946-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR