1-Fluoro-4-iodobenzene, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 4-Fluoroiodobenzene

MQ100

219428-50G 219428-5G
SIAL219428-50GEA 114 EUR
SIAL219428-50G SIAL219428-5G
1-Fluoro-4-iodobenzene, Sigma-Aldrich®
1-Fluoro-4-iodobenzene
Formula: C₆H₄FI
MW: 222 g/mol
Boiling Pt: 182
Melting Pt: –18 °C
Density: 1.9253 g/cm³ (15 °C)
MDL Number: MFCD00001052
CAS Number: 352-34-1
EINECS: 206-522-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR