2,4-Difluorophenyl isocyanate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,4-Difluorophenylisocyanate
M-Clarity™ quality level = MQ100
250759-25G 250759-5G
SIAL250759-25GEA 67.4 EUR
SIAL250759-25G SIAL250759-5G
2,4-Difluorophenyl isocyanate, Sigma-Aldrich®
2,4-Difluorophenyl isocyanate
Formula: C₇H₃F₂NO
MW: 155.1 g/mol
Boiling Pt: 42 °C (6 mmHg)
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001997
CAS Number: 59025-55-7
UN: 3080
ADR: 6.1,II