2,6-Difluorophenyl isothiocyanate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,6-Difluorophenyl isocyanate, 2,6-Difluorophenylisothiocyanate, 1,3-Difluoro-2-isothiocyanatobenzene
M-Clarity™ quality level = MQ100
308196-1G 308196-5G
SIAL308196-1GEA 54.9 EUR
SIAL308196-1G SIAL308196-5G
2,6-Difluorophenyl isothiocyanate, Sigma-Aldrich®
2,6-Difluorophenyl isothiocyanate
Formula: C₇H₃F₂NS
MW: 171.17 g/mol
Boiling Pt: 106…107 °C (35 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD00041047
CAS Number: 207974-17-2
UN: 2930
ADR: 6.1,II