3-Fluorophenyl isothiocyanate
Supplier: Thermo Scientific
TL00416.1 TL00416.10 TL00416.50
MAYBTL00416.1EA 0 EUR
MAYBTL00416.1 MAYBTL00416.10 MAYBTL00416.50
3-Fluorophenyl isothiocyanate
3-Fluorophenyl isothiocyanate
Formula: C₇H₄FNS
Boiling Pt: 226…227 °C (1013 hPa)
Density: 1.27 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004804
CAS Number: 404-72-8
EINECS: 206-967-7
UN: 2922
ADR: 8,III